Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007617483.1 | XP_007615673.1 | 4 - 5 | 707 | 81 | |
GenBank mRNA | JI884580.1 | - | 4 - 5 | 658 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |