Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ninj2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641061.1 | XP_007639251.1 | 3 - 4 | 462 | 105 | |
GenBank mRNA | JI889534.1 | - | 2 - 3 | 209 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |