Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vsig10l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653251.1 | XP_007651441.1 | 3 - 4 | 1160 | 83 |