Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506720.2 | XP_003506768.1 | 2 - 3 | 476 | 76 | |
GenBank mRNA | JI862495.1 | - | 2 - 3 | 463 | 76 | |
JP048674.1 | - | 2 - 3 | 463 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Probe spans exons |