Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fosl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650039.1 | XP_007648229.1 | 2 - 3 | 551 | 93 | |
GenBank mRNA | JI867458.1 | - | 1 - 2 | 141 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |