Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnttip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513756.2 | XP_003513804.1 | 2 - 3 | 1724 | 111 | |
GenBank mRNA | JI893431.1 | - | 2 - 3 | 1805 | 111 | |
JP057327.1 | - | 2 - 3 | 1805 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |