Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lekr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498405.2 | XP_003498453.1 | 8 - 9 | 1298 | 96 | |
XM_007627369.1 | XP_007625559.1 | 9 - 10 | 1312 | 96 | ||
XM_007627377.1 | XP_007625567.1 | 8 - 9 | 1153 | 96 |