Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lekr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498405.2 | XP_003498453.1 | 9 - 10 | 1390 | 83 | |
XM_007627369.1 | XP_007625559.1 | 10 - 11 | 1404 | 83 | ||
XM_007627377.1 | XP_007625567.1 | 9 - 10 | 1245 | 83 |