Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100763131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509581.2 | XP_003509629.1 | 3 - 4 | 283 | 93 | |
GenBank mRNA | JP045829.1 | - | 2 - 3 | 192 | 93 |