Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angptl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640545.1 | XP_007638735.1 | 4 - 5 | 415 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |