Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ascl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648724.1 | XP_007646914.1 | 7 - 8 | 1544 | 81 | |
GenBank mRNA | JI874104.1 | - | 1 - 2 | 179 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |