Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crhbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498707.2 | XP_003498755.1 | 4 - 5 | 928 | 83 | |
XM_007632075.1 | XP_007630265.1 | 5 - 6 | 807 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |