Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nacc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501926.2 | XP_003501974.1 | 3 - 4 | 1145 | 65 | |
XM_007642543.1 | XP_007640733.1 | 4 - 5 | 1216 | 65 | ||
GenBank mRNA | JI874796.1 | - | 2 - 3 | 514 | 65 | |
JP047276.1 | - | 2 - 3 | 514 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |