Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504745.2 | XP_003504793.1 | 3 - 4 | 263 | 67 | |
GenBank mRNA | JP058037.1 | - | 2 - 3 | 162 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |