Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin28a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509731.1 | XP_003509779.1 | 3 - 4 | 392 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |