Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Triml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512585.1 | XP_003512633.1 | 3 - 4 | 478 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |