Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007646799.1 | XP_007644989.1 | 22 - 23 | 3180 | 52 | |
XM_007646800.1 | XP_007644990.1 | 21 - 22 | 3117 | 52 | ||
XM_007646801.1 | XP_007644991.1 | 10 - 11 | 1220 | 52 | ||
GenBank mRNA | JI890324.1 | - | 4 - 5 | 452 | 52 | |
JP043903.1 | - | 4 - 5 | 452 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |