Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tprg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495600.2 | XP_003495648.2 | 2 - 3 | 347 | 73 | |
XM_007642472.1 | XP_007640662.1 | 3 - 4 | 352 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |