Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498466.2 | XP_003498514.2 | 2 - 3 | 300 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |