Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt61a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498782.2 | XP_003498830.1 | 2 - 3 | 441 | 77 | |
GenBank mRNA | JI872039.1 | - | 2 - 3 | 429 | 77 | |
JP048466.1 | - | 2 - 3 | 429 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Probe spans exons |