Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrdc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642449.1 | XP_007640639.1 | 1 - 2 | 248 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |