Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh3b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650570.1 | XP_007648760.1 | 5 - 6 | 768 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |