Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aen | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512538.2 | XP_003512586.2 | 1 - 2 | 685 | 80 | |
GenBank mRNA | JI868882.1 | - | 1 - 2 | 685 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |