Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007609832.1 | XP_007608022.1 | 4 - 5 | 557 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |