Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651201.1 | XP_007649391.1 | 3 - 4 | 642 | 92 | |
GenBank mRNA | JP057858.1 | - | 3 - 4 | 722 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |