Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007622640.1 | XP_007620830.1 | 3 - 4 | 1961 | 61 | |
GenBank mRNA | JI871363.1 | - | 1 - 2 | 1255 | 61 | |
JP052846.1 | - | 1 - 2 | 1255 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |