Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc28a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496542.2 | XP_003496590.1 | 3 - 4 | 699 | 86 | |
XM_007651121.1 | XP_007649311.1 | 3 - 4 | 699 | 86 | ||
GenBank mRNA | JP051965.1 | - | 3 - 4 | 489 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |