Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499422.2 | XP_003499470.1 | 3 - 4 | 296 | 74 | |
XM_007640181.1 | XP_007638371.1 | 3 - 4 | 195 | 74 | ||
GenBank mRNA | JI862042.1 | - | 3 - 4 | 296 | 74 | |
JP052013.1 | - | 3 - 4 | 296 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |