Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam81b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500007.1 | XP_003500055.1 | 5 - 6 | 678 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |