Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653796.1 | XP_007651986.1 | 9 - 10 | 2402 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |