Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504978.2 | XP_003505026.1 | 1 - 2 | 680 | 84 | |
GenBank mRNA | JI866413.1 | - | 1 - 2 | 87 | 84 | |
JP050038.1 | - | 1 - 2 | 87 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |