Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499436.2 | XP_003499484.1 | 4 - 5 | 662 | 87 | |
XM_007640207.1 | XP_007638397.1 | 4 - 5 | 701 | 87 | ||
GenBank mRNA | JP044159.1 | - | 4 - 5 | 611 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |