Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501652.2 | XP_003501700.2 | 14 - 15 | 2271 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |