Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsc22d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510626.2 | XP_003510674.1 | 2 - 3 | 1518 | 112 | |
XM_007651249.1 | XP_007649439.1 | 2 - 3 | 1556 | 112 | ||
GenBank mRNA | JI877936.1 | - | 1 - 2 | 108 | 112 | |
JP040514.1 | - | 1 - 2 | 108 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |