Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsc22d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510626.2 | XP_003510674.1 | 3 - 4 | 1679 | 80 | |
XM_007651249.1 | XP_007649439.1 | 3 - 4 | 1717 | 80 | ||
GenBank mRNA | JI877936.1 | - | 2 - 3 | 269 | 80 | |
JP040514.1 | - | 2 - 3 | 269 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |