Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsc22d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510626.2 | XP_003510674.1 | 4 - 5 | 1724 | 69 | |
GenBank mRNA | JI877936.1 | - | 3 - 4 | 314 | 69 | |
JP040514.1 | - | 3 - 4 | 314 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |