Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007652261.1 | XP_007650451.1 | 2 - 3 | 298 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |