Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645208.1 | XP_007643398.1 | 4 - 5 | 1567 | 90 | |
XM_007645209.1 | XP_007643399.1 | 4 - 5 | 1531 | 90 | ||
GenBank mRNA | JI862516.1 | - | 1 - 2 | 225 | 90 | |
JP063661.1 | - | 1 - 2 | 225 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |