Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ca9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505009.2 | XP_003505057.1 | 6 - 7 | 949 | 82 | |
GenBank mRNA | JI868912.1 | - | 1 - 2 | 70 | 82 | |
JP063035.1 | - | 1 - 2 | 70 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |