Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513529.2 | XP_003513577.1 | 1 - 2 | 925 | 79 | |
GenBank mRNA | JI877116.1 | - | 1 - 2 | 964 | 79 | |
JP054182.1 | - | 1 - 2 | 964 | 79 |