Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642261.1 | XP_007640451.1 | 2 - 3 | 1024 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Ion Channel Probe spans exons |