Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007646803.1 | XP_007644993.1 | 1 - 2 | 200 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |