Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653051.1 | XP_007651241.1 | 3 - 4 | 457 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Protein Phosphatase Probe spans exons |