Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653051.1 | XP_007651241.1 | 4 - 5 | 610 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Protein Phosphatase Probe spans exons |