Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500823.2 | XP_003500871.1 | 3 - 4 | 777 | 91 | |
GenBank mRNA | JI863626.1 | - | 2 - 3 | 254 | 91 | |
JP039354.1 | - | 2 - 3 | 254 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |