Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1g3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504676.1 | XP_003504724.1 | 2 - 3 | 179 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Transporter 3' Most Probe spans exons |