Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508532.1 | XP_003508580.1 | 8 - 9 | 1068 | 103 | |
XM_003508534.1 | XP_003508582.1 | 8 - 9 | 1068 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |