Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650945.1 | XP_007649135.1 | 4 - 5 | 1533 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |