Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495484.1 | XP_003495532.1 | 4 - 5 | 2076 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |