Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498584.2 | XP_003498632.1 | 3 - 4 | 303 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |